Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucose 6 Phosphate Dehydrogenase Antibody (2F6), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB310131 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310131 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310131 Supplier Novus Biologicals Supplier No. NBP130358MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glucose 6 Phosphate Dehydrogenase Monoclonal antibody specifically detects Glucose 6 Phosphate Dehydrogenase in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Glucose 6 Phosphate Dehydrogenase
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 2F6
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias G6PD, G6PD1EC 1.1.1.49, glucose-6-phosphate dehydrogenase
Host Species Mouse
Immunogen Recombinant human Glucose 6 Phosphate Dehydrogenase (35-506aa) purified from E. coli
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 2539
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.