Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucose 6 Phosphate Dehydrogenase Antibody (2H7), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB222125AFK Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB222125AFK 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB222125AFK Supplier Novus Biologicals Supplier No. NBP222125AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glucose 6 Phosphate Dehydrogenase Monoclonal specifically detects Glucose 6 Phosphate Dehydrogenase in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Glucose 6 Phosphate Dehydrogenase
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2H7
Conjugate Alexa Fluor 488
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias G6PD, G6PD1EC 1.1.1.49, glucose-6-phosphate dehydrogenase
Gene Symbols G6PD
Host Species Mouse
Immunogen Purified recombinant fragment of human Glucose 6 Phosphate Dehydrogenase expressed in E. coli. [Uniprot: P11413]
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 2539
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.