Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucose 6 Phosphate Dehydrogenase Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB100236646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100236646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100236646 Supplier Novus Biologicals Supplier No. NB100236JF646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Glucose6PhosphateDehydrogenase Polyclonal specifically detects Glucose6PhosphateDehydrogenase in Human, Mouse samples. It is validated for Immunoblot, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot.

Specifications

Antigen Glucose6PhosphateDehydrogenase
Applications Immunoblot, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen, Immunoblotting
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias G6PD, G6PD1EC 1.1.1.49, glucose-6-phosphate dehydrogenase
Gene Symbols G6PD
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 50 and 100 of human Glucose-6-Phosphate Dehydrogenase using the numbering given in entry NP_000393.2 (GeneID 2539).
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Cellular Markers, Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 2539
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.