Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pointe Scientific Glucose Dry Powder Reagents
Shop All Pointe Scientific Products
Click to view available options
Quantity:
10 x 100mL (3.4 oz.)
1000 mL
5 x 600 mL
Form:
Dry Powder
Liquid
Formulation:
Glucose Oxidase >15U/mL, Peroxidase (horseradish) 800U/L, 4-Aminoantipyrine >15U/mL, Phosphate Buffer pH 7.5 ± 0.1, Sodium p-Hydroxybenzoate, non-reactive Stabilizers and fillers, Sodium Azide 0.1%
Glucose Oxidase (microbial) 12000U/L, Peroxidase (horseradish) >1000U/L,4-AAP >0.3mM, Phenol 4mM, Buffer pH 7.4 ± 0.1, non-reactive Stabilizers
pH:
7.3
7.4
Description
Includes
- Glucose Oxidase >15u/ml; Peroxidase (horseradish) 800U/L; 4-
- Aminoantipyrine 0.38mM; Phosphate Buffer; pH 7.5±0.1; Sodium p-Hydroxybenzoate 5mM; non-reactive Stabilizers and fillers; Sodium Azide 0.1%
- Method: Oxidase/Trinder, Endpoint
- Format: Dry powder
- Wavelength: 500nm
- Linearity: 500mg/dL
- Expected Values: 70 to 105mg/dL
- Reconstituted stability: 30 days at 2 to 8°C
For the quantitative determination of glucose in serum
Specifications
Specifications
| Certifications/Compliance | CE Marked |
| Type | Glucose (Oxidase) Reagent Set |
| Formulation | Glucose Oxidase >15U/mL, Peroxidase (horseradish) 800U/L, 4-Aminoantipyrine >15U/mL, Phosphate Buffer pH 7.5 ± 0.1, Sodium p-Hydroxybenzoate, non-reactive Stabilizers and fillers, Sodium Azide 0.1% |
| For Use With (Application) | Quantitative determination of Glucose in Serum, in-vitro diagnostic use only |
| pH | 7.4 |
| Quantity | 1000 mL |
| Form | Dry Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction