Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucose Oxidase Antibody, Novus Biologicals™
SDP

Catalog No. NBP24418650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP24418650 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP24418650 Supplier Novus Biologicals Supplier No. NBP24418650UG
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Glucose Oxidase Polyclonal specifically detects Glucose Oxidase in Fungi samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen Glucose Oxidase
Applications Western Blot, ELISA, Immunohistochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000 - 1:5000, ELISA 1:2000 - 1:10000, Immunohistochemistry 1:10-1:500
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% sodium azide
Gene Alias Beta D Glucose Oxygen 1 Oxido Reductase, Glucose Oxyhydrase, GOD
Host Species Goat
Immunogen Glucose Oxidase [Aspergillus niger]. Uniprot ID P13006
Quantity 50 μg
Primary or Secondary Primary
Target Species Fungi
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.