Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucosylceramidase/GBA Antibody (OTI4G4), Novus Biologicals™
SDP

Catalog No. NBP245829 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP245829 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP245829 Supplier Novus Biologicals Supplier No. NBP245829
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot, Immunohistochemistry

Glucosylceramidase/GBA Monoclonal specifically detects Glucosylceramidase/GBA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Glucosylceramidase/GBA
Applications Immunohistochemistry (Paraffin), Immunohistochemistry
Classification Monoclonal
Clone OTI4G4
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry 1:150, Immunohistochemistry-Paraffin 1:150
Formulation PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
Gene Alias Acid beta-glucosidase, alglucerase, Beta-glucocerebrosidase, D-glucosyl-N-acylsphingosine glucohydrolase, EC 3.2.1.45, GBA1, GC, GCB, GLUC, glucosidase, beta, acid, glucosidase, beta; acid (includes glucosylceramidase), glucosylceramidase, imiglucerase, lysosomal glucocerebrosidase
Gene Symbols GBA
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 40-315 of human GBA (NP_000148) produced in E.coli.
Molecular Weight of Antigen 55.5 kDa
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2629
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.