Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glucosylceramidase/GBA Rabbit anti-Human, Mouse, Rat, Clone: JM10-76, Novus Biologicals™
SDP

Catalog No. NBP266871 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266871 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266871 Supplier Novus Biologicals Supplier No. NBP266871
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Glucosylceramidase/GBA Monoclonal specifically detects Glucosylceramidase/GBA in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Glucosylceramidase/GBA
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JM10-76
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias Acid beta-glucosidase, alglucerase, Beta-glucocerebrosidase, D-glucosyl-N-acylsphingosine glucohydrolase, EC 3.2.1.45, GBA1, GC, GCB, GLUC, glucosidase, beta, acid, glucosidase, beta; acid (includes glucosylceramidase), glucosylceramidase, imiglucerase, lysosomal glucocerebrosidase
Gene Symbols GBA
Host Species Rabbit
Immunogen Synthetic peptide within Human Glucosylceramidase/GBA aa 477-534/ 536. (SwissProt: P04062 Human; SwissProt: P17439 Mouse; Entrez Gene:684536 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2629
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.