Learn More
Invitrogen™ GluR1 Monoclonal Antibody (N355/1), FITC

Description
1 μg/mL of MA5-45628 was sufficient for detection of GluA1/GluR1 in 20 μg of mouse brain membrane lysate and assayed by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 100kDa. Does not cross-react with GluR2. This antibody was formerly sold as clone S355-1.
Specifications
Specifications
| Antigen | GluR1 |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | N355/1 |
| Concentration | 1 mg/mL |
| Conjugate | FITC |
| Formulation | PBS, 9.09mM sodium bicarbonate with 640.91mM DMSO, 136.36mM ethanolamine and no preservative; pH 7.4 |
| Gene | GRIA1 |
| Gene Accession No. | P19490, P23818, P42261 |
| Gene Alias | 2900051M01Rik; AI853806; AMPA 1; AMPA glutamate receptor; AMPA receptor GluR1/A; AMPA-selective glutamate receptor 1; Glr1; Glr-1; GluA1; GLUH 1; GLUH1; GluR K1; GLUR1; Glur-1; GluR1 flip; GluR1 flop; GLURA; GluR-A; GluRK1; gluR-K1; glutamate ionotropic receptor AMPA type subunit 1; glutamate receptor; glutamate receptor 1; glutamate receptor A; glutamate receptor ionotropic, AMPA 1; glutamate receptor subunit GluR1; glutamate receptor, ionotropic, AMPA 1; glutamate receptor, ionotropic, AMPA1 (alpha 1); GPRC1A; Gria 1; Gria1; GRM1A; HBGR1; HGBR1; HIPA; HIPA1; MGC13325; MGC133252; NMDA1; NMDAR1; NR1; OTTHUMP00000160643; OTTHUMP00000165781; OTTHUMP00000224241; OTTHUMP00000224242; OTTHUMP00000224243; RP11-350O14.1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.