Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GluR1, p Ser831 Antibody, Novus Biologicals™
SDP

Catalog No. NB11093482 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11093482 0.15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11093482 Supplier Novus Biologicals Supplier No. NB11093482
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GluR1 Polyclonal specifically detects GluR1 in Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen GluR1 (p Ser831)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative
Gene Accession No. P23818, P19490
Gene Alias AMPA 1, AMPA-selective glutamate receptor 1, GluA1, GLUH1, GluR-1, GLUR1gluR-A, GluR-A, GLURA, GluR-K1, glutamate receptor 1, Glutamate receptor ionotropic, AMPA 1, glutamate receptor, ionotropic, AMPA 1, HBGR1, MGC133252
Gene Symbols GRIA1
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser831 conjugated to KLH. Accession # P19490
Molecular Weight of Antigen 100 kDa
Purification Method Antigen Affinity-purified
Quantity 0.15 mL
Regulatory Status RUO
Research Discipline Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 2890
Test Specificity Specific for the ∼100k GluR1 protein phosphorylated at Ser831. Immunolabeling is blocked by the phosphopeptide used as antigen but not by the corresponding dephosphopeptide. Immunolabeling is completely eliminated by lambda-phosphatase treatment.
Target Species Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.