Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GluR2/3 Antibody, Novus Biologicals™
SDP

Catalog No. NB300302 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300302 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300302 Supplier Novus Biologicals Supplier No. NB300302
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GluR2/3 Polyclonal specifically detects GluR2/3 in Rat samples. It is validated for Western Blot.

Specifications

Antigen GluR2/3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative
Gene Accession No. P19491
Gene Alias AMPA 2, glutamate receptor, ionotropic, AMPA 2
Gene Symbols GRIA2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acid residues from the C-terminal region conjugated to KLH. Accession # P19491
Molecular Weight of Antigen 100 kDa
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Tumor Suppressors, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2891
Test Specificity Specific for the ∼100k GluR2/3 protein.
Target Species Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.