Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GluR2/3 Rabbit anti-Mouse, Rat, Clone: 4K7G5, Novus Biologicals™
SDP

Catalog No. NB167384 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167384 20 μg
NB167383 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167384 Supplier Novus Biologicals Supplier No. NBP31614620UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

GluR2/3 Monoclonal antibody specifically detects GluR2/3 in Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen GluR2/3
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 4K7G5
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias AMPA 2, glutamate receptor, ionotropic, AMPA 2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 784-883 of human GluR2/3 (P42262).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Tumor Suppressors, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2891
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.