Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GluR2 Antibody (3A11) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB019445 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019445 0.025 mg
NBP229731 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019445 Supplier Novus Biologicals Supplier No. NBP2297310.025MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GluR2 Monoclonal specifically detects GluR2 in Human, Rat, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Radioimmunoassay.

Specifications

Antigen GluR2
Applications Immunoprecipitation, Radioimmune Assays (RIA), Immunofluorescence, Western Blot, Immunohistochemistry, Immunocytochemistry, ELISA
Classification Monoclonal
Clone 3A11
Concentration 1.7 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:500-1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoprecipitation 1:10-1:500, Radioimmunoassay 1:100-1:2000
Formulation PBS with No Preservative
Gene Accession No. Q14416
Gene Alias AMPA 2, glutamate receptor, ionotropic, AMPA 2
Gene Symbols GRIA2
Host Species Mouse
Immunogen Recombinant fusion protein TrpE-GluR2.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Tumor Suppressors, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2891
Test Specificity Glutamate Receptor 2 (GluR2) by Western Blot and RIA. When using MAB396 for immunohistochemistry or immunoprecipitation it recognizes both GluR2 and GluR4.
Target Species Rat, Primate, Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.