Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GluR7/GRIK3 Antibody (2B4H1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP261783
Description
GluR7/GRIK3 Monoclonal specifically detects GluR7/GRIK3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
GluR7/GRIK3 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
EAA5, Excitatory amino acid receptor 5, GLR7, GluR7, GluR-7, GluR7a, GLUR7dJ1090M5.1 (glutamate receptor, ionotropic, kainate 3 (GLUR7)), Glutamate receptor 7, glutamate receptor, ionotropic kainate 3, glutamate receptor, ionotropic, kainate 3 | |
Mouse | |
104 kDa | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, ELISA | |
2B4H1 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
GRIK3 | |
Purified recombinant fragment of human GRIK3 (AA: extra 32-173) expressed in E. Coli. | |
Protein G purified | |
Neuronal Cell Markers, Neurotransmission | |
2899 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction