Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GluR7/GRIK3 Antibody (2B4H1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | GluR7/GRIK3 |
---|---|
Clone | 2B4H1 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
Applications | Western Blot, Flow Cytometry, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61783
![]() |
Novus Biologicals
NBP261783 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
GluR7/GRIK3 Monoclonal specifically detects GluR7/GRIK3 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
GluR7/GRIK3 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Mouse | |
EAA5, Excitatory amino acid receptor 5, GLR7, GluR7, GluR-7, GluR7a, GLUR7dJ1090M5.1 (glutamate receptor, ionotropic, kainate 3 (GLUR7)), Glutamate receptor 7, glutamate receptor, ionotropic kainate 3, glutamate receptor, ionotropic, kainate 3 | |
GRIK3 | |
IgG1 | |
Protein G purified |
2B4H1 | |
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
Neuronal Cell Markers, Neurotransmission | |
2899 | |
Purified recombinant fragment of human GRIK3 (AA: extra 32-173) expressed in E. Coli. | |
Primary | |
104 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title