Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glut1 Antibody (rGLUT1/2476), DyLight 550, Novus Biologicals™
SDP

Catalog No. NB392214 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392214 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392214 Supplier Novus Biologicals Supplier No. NBP308479R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glut1 Monoclonal antibody specifically detects Glut1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Glut1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone rGLUT1/2476
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias Choreoathetosis/Spasticity, Episodic (Paroxysmal Choreoathetosis), CSE, DYT17, DYT18, DYT9, EIG12, Glucose transporter type 1, erythrocyte/brain, GLUT-1, GLUT1DS, HepG2 glucose transporter, HTLVR, Human T-Cell Leukemia Virus (I and II) Receptor, MGC141895, MGC141896, PED, solute carrier family 2 (facilitated glucose transporter), member 1, Solute Carrier Family 2 Member 1, solute carrier family 2, facilitated glucose transporter member 1
Host Species Mouse
Immunogen Recombinant fragment of human Glut1 protein (around aa 203-305) (exact sequence is proprietary) (Uniprot: P11166)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Cellular Markers, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroscience, Plasma Membrane Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6513
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.