Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLUT10 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB000940 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB000940 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB000940 Supplier Novus Biologicals Supplier No. NB100490SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GLUT10 Polyclonal specifically detects GLUT10 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen GLUT10
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000
Formulation PBS with 0.02% Sodium Azide
Gene Alias ATS, Glucose transporter type 10, GLUT-10, GLUT10MGC126706, solute carrier family 2 (facilitated glucose transporter), member 10, solute carrier family 2, facilitated glucose transporter member 10
Gene Symbols SLC2A10
Host Species Rabbit
Immunogen A synthetic peptide containing residues 367-385 (ILSTAKKTKPHPRSGDPSA) of the human GLUT10 protein.
Molecular Weight of Antigen 52 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Research Discipline Diabetes Research
Primary or Secondary Primary
Gene ID (Entrez) 81031
Test Specificity Human GLUT10 protein.
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.