Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLUT12 Antibody, Novus Biologicals™
SDP

Catalog No. NB186413 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB186413 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB186413 Supplier Novus Biologicals Supplier No. NBP312263
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GLUT12 Polyclonal antibody specifically detects GLUT12 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Dot Blot

Specifications

Antigen GLUT12
Applications Western Blot, ELISA, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Dot Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA 1:4500, Immunocytochemistry/Immunofluorescence 1:200, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin, Dot Blot 1:4500
Formulation Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide
Gene Alias GLUT-12, GLUT12GLUT8Glucose transporter type 12, solute carrier family 2 (facilitated glucose transporter), member 12, solute carrier family 2, facilitated glucose transporter member 12
Host Species Rabbit
Immunogen Synthetic peptide from the C-terminus of Rat GLUT12.
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154091
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.