Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glut2 Antibody (199017), HRP, Novus Biologicals™
SDP

Catalog No. NB175162 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175162 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175162 Supplier Novus Biologicals Supplier No. FAB1414H
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Glut2 Monoclonal specifically detects Glut2 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen Glut2
Applications CyTOF, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 199017
Conjugate HRP
Formulation PBS
Gene Alias GLUT-2, GLUT2 Glucose transporter type 2, liver, SLC2A2, solute carrier family 2 (facilitated glucose transporter), member 2, solute carrier family 2, facilitated glucose transporter member 2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Glut2, Met1-Val524, Accession # P11168
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Stem Cell Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 6514
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.