Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glut3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP236485 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP236485 100 μL
NB019673 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP236485 Supplier Novus Biologicals Supplier No. NBP236485
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Glut3 Polyclonal specifically detects Glut3 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Glut3
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:2000-1:5000
Formulation PBS pH 7.4, 40% Glycerol. with 0.05% Sodium Azide
Gene Accession No. P11169
Gene Alias FLJ90380, GLUT-3, GLUT3Glucose transporter type 3, brain, solute carrier family 2 (facilitated glucose transporter), member 3, solute carrier family 2, facilitated glucose transporter member 3
Gene Symbols SLC2A3
Host Species Rabbit
Immunogen Raised against Peptide-KLH. Synthetic peptide corresponding to a region derived from 466-479 amino acids of human Glut3.
Molecular Weight of Antigen 54 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6515
Test Specificity Detects endogenous level of total Glut3 protein.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.