Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glut3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | Glut3 |
---|---|
Concentration | 0.8 mg/mL |
Dilution | Western Blot 1:500-1:2000, ELISA 1:2000-1:5000 |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Description
Glut3 Polyclonal specifically detects Glut3 in Human samples. It is validated for Western Blot, ELISA.Specifications
Glut3 | |
Western Blot 1:500-1:2000, ELISA 1:2000-1:5000 | |
Polyclonal | |
Rabbit | |
Human | |
FLJ90380, GLUT-3, GLUT3Glucose transporter type 3, brain, solute carrier family 2 (facilitated glucose transporter), member 3, solute carrier family 2, facilitated glucose transporter member 3 | |
SLC2A3 | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
0.8 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
P11169 | |
6515 | |
IgG | |
Affinity Purified | |
54 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title