Learn More
GLUT3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | GLUT3 |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2869
![]() ![]() |
Proteintech Group Inc
204031AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody was generated against the internal region of human GLUT3 and may cross-react with GLUT14.20403-1-AP antibody detects the bands around 48-60 kDa in SDS-PAGE.
Glucose is the major source of our energy and there are numerous isoforms of the glucose transporter in mammals, including Glut1, Glut2, Glut3, Glut4, Glut5, Glut6, Glut7, Glut8 and Glut9. The Glut5 gene located on the short arm of human chromosome 1 encodes a 501-amino acid facilitative glucose transporter. Glut5 mRNA is highly expressed in small intestine and to a lesser extent in kidney, skeletal muscle and adipose tissue. Glut5 plays a critical role in fructose absorption in the small intestine and its expression is highly induced when exposed to a fructose-enriched diet. Glut5 transporter expressed in human skeletal muscle is specifically localized to the plasma membrane, where it participates in regulating hexose transfer across the sarcolemma. Glut8, a novel glucose transporter-like protein, exhibits significant sequence similarity with the other members of sugar transporter family. Glut8 comprises 12 putative membrane-spanning helices and several conserved motifs, which are important for transport activity. In human tissues, Glut8 is predomitly expressed in testis and, to a lesser extent, in most other tissues including skeletal muscle, heart, small intestine and brain.Specifications
GLUT3 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P11169, P32037 | |
20527, 6515 | |
GLUT3 Fusion Protein Ag14203 | |
Primary | |
-20°C | |
SLC2A3 |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
GLUT 3, GLUT3, SLC2A3 | |
SLC2A3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.