Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glut4 Antibody (925932), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB310679 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310679 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310679 Supplier Novus Biologicals Supplier No. FAB86541APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glut4 Monoclonal specifically detects Glut4 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Glut4
Applications Flow Cytometry
Classification Monoclonal
Clone 925932
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias GLUT-4, GLUT4Glucose transporter type 4, insulin-responsive, insulin-responsive glucose transporter type 4, solute carrier family 2 (facilitated glucose transporter), member 4, solute carrier family 2, facilitated glucose transporter member 4
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Glut4, Met1-Asp509, Accession # P14672
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Core ESC Like Genes, Diabetes Research, Hematopoietic Stem Cell Markers, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroendocrinology, Neuronal Cell Markers, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6517
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.