Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glut4 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP14933A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP14933A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP14933A94 Supplier Novus Biologicals Supplier No. NBP149533AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Glut4 Polyclonal specifically detects Glut4 in Human, Mouse, Rat, Plant samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), Knockdown Validated.

Specifications

Antigen Glut4
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), KnockDown Validated
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias GLUT-4, GLUT4Glucose transporter type 4, insulin-responsive, insulin-responsive glucose transporter type 4, solute carrier family 2 (facilitated glucose transporter), member 4, solute carrier family 2, facilitated glucose transporter member 4
Gene Symbols SLC2A4
Host Species Rabbit
Immunogen A synthetic peptide from the human Glucose Transporter GLUT4 protein [UniProt P14672]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6517
Target Species Human, Mouse, Rat, Plant
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.