Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glut4 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP149533G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149533G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149533G Supplier Novus Biologicals Supplier No. NBP149533G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Glut4 Polyclonal specifically detects Glut4 in Human, Mouse, Rat, Plant samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), Knockdown Validated.

Specifications

Antigen Glut4
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular), Knockdown Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P14672
Gene Alias GLUT-4, GLUT4Glucose transporter type 4, insulin-responsive, insulin-responsive glucose transporter type 4, solute carrier family 2 (facilitated glucose transporter), member 4, solute carrier family 2, facilitated glucose transporter member 4
Gene Symbols SLC2A4
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human Glucose Transporter GLUT4 protein (between residues 480-509) [UniProt P14672]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones
Primary or Secondary Primary
Gene ID (Entrez) 6517
Test Specificity Human and mouse. Predicted to react with rat based on 100% sequence homology.
Target Species Human, Mouse, Rat, Plant
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.