Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glut5 Antibody (195205), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB175111 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175111 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175111 Supplier Novus Biologicals Supplier No. FAB1349J
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Glut5 Monoclonal specifically detects Glut5 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen Glut5
Applications CyTOF, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 195205
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias Fructose transporter, Glucose transporter type 5, small intestine, GLUT-5, GLUT5glucose transporter-like protein 5, solute carrier family 2 (facilitated glucose transporter), member 5, solute carrier family 2 (facilitated glucose/fructose transporter), member 5, solute carrier family 2, facilitated glucose transporter member 5
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Glut5, Met1-Gln501, Accession # P22732
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6518
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.