Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLUT9 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP106565UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP106565UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP106565UV Supplier Novus Biologicals Supplier No. NBP106565UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GLUT9 Polyclonal specifically detects GLUT9 in Human, Mouse, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GLUT9
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias facilitated glucose transporter member 9, GLUT-9, GLUT9Glut9, GLUTX, solute carrier family 2 (facilitated glucose transporter), member 9, UAQTL2, urate voltage-driven efflux transporter 1
Gene Symbols SLC2A9
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human GLUT9 (within residues 10-60). [Swiss-ProtNo. Q9NRM0]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 56606
Test Specificity This antibody is specific for the human GLUT9 protein Isoform 1 (SLC2A9-L).
Target Species Human, Mouse, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.