Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutamate Antibody (S1), Novus Biologicals™
SDP

Catalog No. NBP229884 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229884 0.1 mL
NB019499 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP229884 Supplier Novus Biologicals Supplier No. NBP229884

Mouse Monoclonal Antibody

Glutamate Monoclonal specifically detects Glutamate in Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen Glutamate
Applications Immunohistochemistry
Classification Monoclonal
Clone S1
Conjugate Unconjugated
Dilution Immunohistochemistry 1:500-1:2500
Host Species Mouse
Immunogen Glutamate-Glutaraldehyde-BSA.
Purification Method Ion exchange chromatography
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Glutamate The cross-reactivities were determined using an ELISA test by competition experiments with the following compounds: Compound Cross-reactivity Glutamate-G-BSA 1 Aspartate-G-BSA 1/100,000 GABA-G-BSA 1/100,000 Glutamate 1/100,000 Abbreviations: (G) Glutaraldehyde (BSA) Bovine Serum Albumin NOTE: Antibody reactivity REQUIRES glutaraldehyde fixation thus some glutaraldehyde (0.5%-2.0%) needs to be included in the tissue fixation procedure inorder for the proper reactivity.
Target Species Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.