Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutamate Receptor 4 Antibody (1059705), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB297366 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297366 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB297366 Supplier Novus Biologicals Supplier No. FAB113631I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glutamate Receptor 4 Monoclonal antibody specifically detects Glutamate Receptor 4 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry

Specifications

Antigen Glutamate Receptor 4
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 1059705
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate
Gene Alias AMPA-selective glutamate receptor 4, GluA4, GluR4, GluR-4, GLUR4C, GLUR4gluR-D, GluR-D, GLURD, glutamate receptor 4, Glutamate receptor ionotropic, AMPA 4, glutamate receptor, ionotrophic, AMPA 4
Host Species Mouse
Immunogen Synthetic peptide, Accession # P48058
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, GPCR, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2893
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.