Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutamate Receptor 6 Antibody (818235), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB302367 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB302367 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB302367 Supplier Novus Biologicals Supplier No. FAB9610MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glutamate Receptor 6 Monoclonal specifically detects Glutamate Receptor 6 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Glutamate Receptor 6
Applications Flow Cytometry
Classification Monoclonal
Clone 818235
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias EAA4, glutamate receptor 6, glutamate receptor, ionotropic, kainate 2, ionotropic kainate 2, MGC74427
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line CHO-derived human GluR6/GRIK2 protein, Thr32 - Asn549, Accession # Q13002
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, GPCR, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2898
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.