Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutamine Synthetase Antibody (8D7), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB162914 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB162914 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB162914 Supplier Novus Biologicals Supplier No. NBP260240AF532
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Glutamine Synthetase Monoclonal antibody specifically detects Glutamine Synthetase in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen Glutamine Synthetase
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone 8D7
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias cell proliferation-inducing protein 59, EC 4.1.1.15, EC 6.3.1.2, GLNSPIG59, Glutamate decarboxylase, glutamate-ammonia ligase, Glutamate--ammonia ligase, glutamate-ammonia ligase (glutamine synthase), glutamine synthase, glutamine synthetase, GS, PIG43, proliferation-inducing protein 43
Host Species Mouse
Immunogen Recombinant human Glutamine Synthetase (1-373aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Hypoxia, Neurodegeneration, Neuroscience, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2752
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.