Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutamine Synthetase Antibody (OTI1F4), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP270834A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP270834A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP270834A94 Supplier Novus Biologicals Supplier No. NBP270834AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glutamine Synthetase Monoclonal specifically detects Glutamine Synthetase in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Glutamine Synthetase
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1F4
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias cell proliferation-inducing protein 59, EC 4.1.1.15, EC 6.3.1.2, GLNSPIG59, Glutamate decarboxylase, glutamate-ammonia ligase, Glutamate--ammonia ligase, glutamate-ammonia ligase (glutamine synthase), glutamine synthase, glutamine synthetase, GS, PIG43, proliferation-inducing protein 43
Gene Symbols GLUL
Host Species Mouse
Immunogen Full length recombinant protein of human Glutamine Synthetase (NP_002056) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2752
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.