Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutaredoxin 1/GLRX1 Rabbit anti-Human, Alexa Fluor 647, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB087169 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB087169 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB087169 Supplier Novus Biologicals Supplier No. NBP300300AF647

Rabbit Polyclonal Antibody

Glutaredoxin 1/GLRX1 Polyclonal specifically detects Glutaredoxin 1/GLRX1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Glutaredoxin 1/GLRX1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50 mM Sodium Borate
Gene Alias glutaredoxin (thioltransferase), glutaredoxin-1, GRXGRX1, MGC117407, Thioltransferase-1, TTase-1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Glutaredoxin 1/GLRX1 (P35754; Met1-Gln106).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cytokine Research, DNA Repair, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 2745
Test Specificity No cross-reactivity in ELISA with: Human cell lysate (293 cell line)
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.