Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutaredoxin 3/GLRX3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220767 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220767 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220767 Supplier Novus Biologicals Supplier No. NBP220767
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Glutaredoxin 3/GLRX3 Polyclonal specifically detects Glutaredoxin 3/GLRX3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen TXNL2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.61mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:10000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. O76003
Gene Alias FLJ11864, GLRX4, glutaredoxin 3, glutaredoxin 4, glutaredoxin-3, GRX3, GRX4, PICOTbA500G10.4, PKC-interacting cousin of thioredoxin, PKCq-interacting protein, PKC-theta-interacting protein, thioredoxin-like 2, Thioredoxin-like protein 2, TXNL2, TXNL3
Gene Symbols GLRX3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human TXNL2. The exact sequence is proprietary.
Molecular Weight of Antigen 37 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10539
Test Specificity Human, Mouse. Species reactivity based on sequence identity: Rat (96%), Xenopus laevis (80%), Bovine (96%).
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.