Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutathione Peroxidase 1/GPX1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10040805 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10040805 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10040805 Supplier Novus Biologicals Supplier No. NB10040805
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Glutathione Peroxidase 1/GPX1 Polyclonal specifically detects Glutathione Peroxidase 1/GPX1 in Human, Pig samples. It is validated for Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot.

Specifications

Antigen Glutathione Peroxidase 1/GPX1
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Immunohistochemistry 3.7 ug/ml, Immunohistochemistry-Paraffin 3.7 ug/ml, Immunohistochemistry-Frozen 20 ug/ml, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_000572.2
Gene Alias Cellular glutathione peroxidase, EC 1.11.1, EC 1.11.1.9, glutathione peroxidase 1, GPx-1, GSHPx-1, GSHPX1, MGC14399, MGC88245
Gene Symbols GPX1
Host Species Goat
Immunogen Peptide with sequence C-REALPAPSDDATA corresponding to internal region according to NP_000572.2.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2876
Test Specificity This antibody is expected to recognize isoform 1 ( NP_000572.2) only.
Target Species Human, Pig
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.