Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutathione Peroxidase 4/GPX4 Antibody (LHM 2), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB392615 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392615 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392615 Supplier Novus Biologicals Supplier No. NBP308253AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glutathione Peroxidase 4/GPX4 Monoclonal antibody specifically detects Glutathione Peroxidase 4/GPX4 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Glutathione Peroxidase 4/GPX4
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LHM 2
Conjugate Alexa Fluor 405
Formulation 50mM Sodium Borate
Gene Alias EC 1.11.1, EC 1.11.1.12, Glutathione peroxidase 4, glutathione peroxidase 4 (phospholipid hydroperoxidase), GPx-4, GSHPx-4, MCSP, PHGPxsnGPx, phospholipid hydroperoxidase, phospholipid hydroperoxide glutathione peroxidase, mitochondrial, snPHGPx, sperm nucleus glutathione peroxidase
Host Species Mouse
Immunogen A 375P cells crude extract
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2879
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.