Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutathione S-Transferase mu 1/GSTM1 Antibody (CPTC-GSTMu1-3), FITC, Novus Biologicals™
SDP

Catalog No. NB382341 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB382341 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB382341 Supplier Novus Biologicals Supplier No. NBP308254F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Glutathione S-Transferase mu 1/GSTM1 Monoclonal antibody specifically detects Glutathione S-Transferase mu 1/GSTM1 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen Glutathione S-Transferase mu 1/GSTM1
Applications Western Blot, Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone CPTC-GSTMu1-3
Conjugate FITC
Formulation PBS
Gene Alias EC 2.5.1.18, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, glutathione S-aryltransferase, glutathione S-transferase M1, glutathione S-transferase mu 1, GST class-mu 1, GST HB subunit 4, GST1, GSTM1-1, GSTM1a-1a, GSTM1b-1b, GTH4, GTM1, H-B, HB subunit 4, MGC26563, MU, MU-1, S-(hydroxyalkyl)glutathione lyase
Host Species Mouse
Immunogen Recombinant human full-length Glutathione S-Transferase mu 1/GSTM1 protein (Uniprot: P09488)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Asthma, Cancer, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 2944
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.