| Antigen |
Glutathione S-Transferase mu 1/GSTM1 |
| Applications |
Western Blot |
| Classification |
Polyclonal |
| Concentration |
0.5 mg/ml |
| Conjugate |
Unconjugated |
| Dilution |
Western Blot 1.0 ug/ml |
| Formulation |
PBS, 2% Sucrose with 0.09% Sodium Azide |
| Gene Accession No. |
P46439 |
| Gene Alias |
EC 2.5.1.18, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, glutathione S-aryltransferase, glutathione S-transferase M1, glutathione S-transferase mu 1, GST class-mu 1, GST HB subunit 4, GST1, GSTM1-1, GSTM1a-1a, GSTM1b-1b, GTH4, GTM1, H-B, HB subunit 4, MGC26563, MU, MU-1, S-(hydroxyalkyl)glutathione lyase |
| Gene Symbols |
GSTM1 |
| Host Species |
Rabbit |
| Immunogen |
Synthetic peptides corresponding to GSTM1(glutathione S-transferase M1) The peptide sequence was selected from the N terminal of GSTM1. Peptide sequence KKYTMGDAPDYDRSQWLNEKFKLGLDFPNLPYLIDGAHKITQSNAILCYI. |
| Purification Method |
Affinity purified |
| Quantity |
100 μL |
| Regulatory Status |
RUO |
| Research Discipline |
Asthma, Cancer, Immunology |
| Primary or Secondary |
Primary |
| Gene ID (Entrez) |
2944 |
| Test Specificity |
This product recognizes Human, Mouse, Rat and Dog. Computational sequence identity: Bovine 100%; Guinea pig 100%; Golden hamster 100%; Long-tailed dwarf hamster 100%; Tetrahymena thermophila SB210 100%; Sarcoptes scabiei type hominis 100%; Crab-eating macaque 100%; Sumatran orangutan 100%; Japanese macaque 100%; Sarcoptes scabiei type suis 100%; Sarcoptes scabiei type canis 100%; Mouse 100%; Dust mite 100%; brown legged grain mite 100%; Rat 100%; Black-legged tick 100%; Caligus clemensi 100%; Blood fluke 100%; Giant panda 100%; Sheep 92%. |
| Reconstitution |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. |
| Target Species |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Goat, Rabbit, Sheep |
| Content And Storage |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Isotype |
IgG |
|
Show More
Show Less
|
|