Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutathione S-transferase Mu 5 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB332378 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB332378 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB332378 Supplier Novus Biologicals Supplier No. NBP297064MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Glutathione S-transferase Mu 5 Polyclonal antibody specifically detects Glutathione S-transferase Mu 5 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen Glutathione S-transferase Mu 5
Applications Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 2.5.1.18, glutathione S-alkyltransferase M5, glutathione S-aralkyltransferase M5, glutathione S-aryltransferase M5, glutathione S-transferase M5, glutathione S-transferase mu 5, GST class-mu 5, GSTM5-5, GTM5, S-(hydroxyalkyl)glutathione lyase M5
Host Species Rabbit
Immunogen Rabbits immunized with purified, recombinant Human Glutathione S-transferase Mu 5 (Accession#: AAH58881.1, Met1-Lys218)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 2949
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.