Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutathione S-Transferase pi 1/GSTP1 Antibody (028), DyLight 350
SDP

Catalog No. NB088469 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB088469 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB088469 Supplier Novus Biologicals Supplier No. NBP289324UV

Rabbit Monoclonal Antibody

Glutathione S-Transferase pi 1/GSTP1 Monoclonal specifically detects Glutathione S-Transferase pi 1/GSTP1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Glutathione S-Transferase pi 1/GSTP1
Applications Western Blot
Classification Monoclonal
Clone 28
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Gene Alias deafness, X-linked 7, EC 2.5.1.18, FAEES3, fatty acid ethyl ester synthase III, glutathione S-transferase P, glutathione S-transferase pi 1, GST class-pi, GST3DFN7, GSTP, GSTP1-1, PI
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the N-terminus of the Human Glutathione S-Transferase pi 1/GSTP1.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, Cancer, Immunology, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2950
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.