Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutathione S-Transferase pi 1/GSTP1 Antibody (8F6), Novus Biologicals™
SDP

Catalog No. NB297896 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB297896 50 μL
NB297897 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB297896 Supplier Novus Biologicals Supplier No. NBP32654050UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Glutathione S-Transferase pi 1/GSTP1 Monoclonal antibody specifically detects Glutathione S-Transferase pi 1/GSTP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen Glutathione S-Transferase pi 1/GSTP1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 8F6
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias deafness, X-linked 7, EC 2.5.1.18, FAEES3, fatty acid ethyl ester synthase III, glutathione S-transferase P, glutathione S-transferase pi 1, GST class-pi, GST3DFN7, GSTP, GSTP1-1, PI
Host Species Rabbit
Immunogen A synthesized peptide derived from Human Glutathione S-Transferase pi 1/GSTP1 [UniProt P09211]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Asthma, Cancer, Immunology, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2950
Target Species Human, Mouse, Rat
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.