Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glutathione S-Transferase pi 1/GSTP1 Antibody (8F6), Novus Biologicals™

Rabbit Monoclonal Antibody
$295.74 - $479.73
Specifications
| Antigen | Glutathione S-Transferase pi 1/GSTP1 |
|---|---|
| Clone | 8F6 |
| Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Classification | Monoclonal |
Description
Glutathione S-Transferase pi 1/GSTP1 Monoclonal antibody specifically detects Glutathione S-Transferase pi 1/GSTP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| Glutathione S-Transferase pi 1/GSTP1 | |
| Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| deafness, X-linked 7, EC 2.5.1.18, FAEES3, fatty acid ethyl ester synthase III, glutathione S-transferase P, glutathione S-transferase pi 1, GST class-pi, GST3DFN7, GSTP, GSTP1-1, PI | |
| A synthesized peptide derived from Human Glutathione S-Transferase pi 1/GSTP1 [UniProt P09211] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 8F6 | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Asthma, Cancer, Immunology, Neuroscience | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 2950 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title