Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glutathione S-Transferase pi 1/GSTP1 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB194465 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB194465 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB194465 Supplier Novus Biologicals Supplier No. NBP176857F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Glutathione S-Transferase pi 1/GSTP1 Polyclonal antibody specifically detects Glutathione S-Transferase pi 1/GSTP1 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Glutathione S-Transferase pi 1/GSTP1
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias deafness, X-linked 7, EC 2.5.1.18, FAEES3, fatty acid ethyl ester synthase III, glutathione S-transferase P, glutathione S-transferase pi 1, GST class-pi, GST3DFN7, GSTP, GSTP1-1, PI
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide from near the center of human GSTP1. The immunogen is located within amino acids 90 - 140 of GSTP1. Amino Acid Squence: YTNYEAGKDDYVKA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Asthma, Cancer, Immunology, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2950
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.