Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLYAT Antibody, Novus Biologicals™
SDP

Catalog No. NBP15471420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15471420 20 μL
NBP154714 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15471420 Supplier Novus Biologicals Supplier No. NBP15471420UL

Rabbit Polyclonal Antibody

GLYAT Polyclonal specifically detects GLYAT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen GLYAT
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q6IB77
Gene Alias ACGNATAAc, Acyl-CoA:glycine N-acyltransferase, Aralkyl acyl-CoA N-acyltransferase, Aralkyl acyl-CoA:amino acid N-acyltransferase, aralkyl-CoA N-acyltransferase, CATEC 2.3.1.13, GATHRP-1(CLP), glycine N-acyltransferase, glycine-N-acyltransferase
Gene Symbols GLYAT
Host Species Rabbit
Immunogen Synthetic peptides corresponding to GLYAT(glycine-N-acyltransferase) The peptide sequence was selected from the N terminal of GLYAT. Peptide sequence HYTNTYQIYSKDPQNCQEFLGSPELINWKQHLQIQSSQPSLNEAIQNLAA.
Molecular Weight of Antigen 34 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10249
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.