Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GLYAT Antibody (OTI4D1), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N271564J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271564J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271564J646 Supplier Novus Biologicals Supplier No. NBP271564JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GLYAT Monoclonal specifically detects GLYAT in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen GLYAT
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4D1
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ACGNATAAc, Acyl-CoA:glycine N-acyltransferase, Aralkyl acyl-CoA N-acyltransferase, Aralkyl acyl-CoA:amino acid N-acyltransferase, aralkyl-CoA N-acyltransferase, CATEC 2.3.1.13, GATHRP-1(CLP), glycine N-acyltransferase, glycine-N-acyltransferase
Gene Symbols GLYAT
Host Species Mouse
Immunogen Full length recombinant protein of human GLYAT (NP_005829) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 10249.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.