Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Glycerin, Synthetic, EP, BP, JP, USP, 99-101%, Spectrum™ Chemical

Click to view available options
Quantity:
500 mL
4 L
4 x 4 L
Packaging:
Amber Glass Bottle
Poly Bottle
Description
Spectrum™ Chemical Glycerin, Synthetic, USP, EP, BP, JP is one of two USP grade Glycerins available from Spectrum with the other being Glycerin, Natural, USP, EP, BP. For those products requiring FDA approvals, if the initial formulation was done with Glycerin, Synthetic, USP, EP, BP, JP, the use of Glycerin, Natural, USP, EP, BP, JP is not interchangeable but would require the new formulation to undergo a new FDA approval process.
Specifications
Specifications
CAS Max % | 100% |
Ignition Residue | 0.01% |
Packaging | Amber Glass Bottle |
Refractive Index | 1.470-1.475 |
Quantity | 4 L |
Specific Gravity | 1.258 |
Percent Purity | 99 to 101% |
Grade | Multi Compendia |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction