Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerin:Water, 3:1, Reagents

Catalog No. 01335167
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-167 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-167 Supplier Reagents Holdings Llc Supplier No. CG0001001A
Only null left
Add to Cart
Add to Cart

Glycerin:Water, 3:1, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 56-81-5 , 7732-18-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym 1,2,3-Propanetriol, Glycerine
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Color Colorless
Boiling Point 121°C
Packaging Poly Bottle
Quantity 1 L
Specific Gravity 1.1994
Grade Reagent
Physical Form Liquid
Chemical Name or Material Glycerin:Water, 3:1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.