Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerol 3 Phosphate Dehydrogenase Antibody, Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB332359 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB332359 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB332359 Supplier Novus Biologicals Supplier No. NBP297069JF549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Glycerol 3 Phosphate Dehydrogenase Polyclonal antibody specifically detects Glycerol 3 Phosphate Dehydrogenase in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen Glycerol 3 Phosphate Dehydrogenase
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias EC 1.1.1, EC 1.1.1.8, FLJ26652, glycerol-3-phosphate dehydrogenase [NAD+], cytoplasmic, glycerol-3-phosphate dehydrogenase 1 (soluble), glycerol-3-phosphate dehydrogenase, soluble, GPD-C, GPDH-C
Host Species Rabbit
Immunogen Rabbits immunized with purified, recombinant Human Glycerol 3 Phosphate Dehydrogenase (Accession#: P21695, Met1-Met349)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2819
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.