Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerol, 99.5+%, for spectroscopy

Catalog No. p-138903
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle

CAS: 56-81-5 | C3H8O3 | 92.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Absorbance (1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.01 max. at 340nm, 0.01 max. at 400nm, 0.02 max. at 320nm, 0.05 max. at 280nm, 0.25 max. at 225nm, 0.7 max. at 210nm, 1 max. at 206nm
Decomposition Information >290°C
Melting Point 18.0°C
Color Black to Brown
Density 1.2610g/mL
Boiling Point 290.0°C
Flash Point 160°C
Assay Percent Range 99.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.4716
Linear Formula HOCH2CH(OH)CH2OH
Quantity 500 mL
Beilstein 01, 502
Specific Gravity 1.261
Merck Index 15, 4520
Solubility Information Solubility in water: >500g/L (20°C). Other solubilities: soluble in methanol, ethanol and 2-propanol, insoluble in ether, benzene, chloroform
Viscosity 1069 mPa.s (20°C)
Vapor Pressure 0.003mbar at 50°C
Formula Weight 92.09
Percent Purity 99.5+%
Grade Spectroscopy
Physical Form Solution
Chemical Name or Material Glycerol, 99.5+%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 200-289-5
RTECSNumber MA8050000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.