Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerol, 99+%, pure, synthetic, ACROS Organics™

Click to view available options
Quantity:
100mL
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
56-81-5
C3H8O3
92.09
PEDCQBHIVMGVHV-UHFFFAOYSA-N
glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
753
CHEBI:17754
propane-1,2,3-triol
C(C(CO)O)O
56-81-5
C3H8O3
92.09
PEDCQBHIVMGVHV-UHFFFAOYSA-N
glycerol,glycerin,glycerine,1,2,3-propanetriol,glycyl alcohol,trihydroxypropane,glyceritol,propanetriol,1,2,3-trihydroxypropane,osmoglyn
753
CHEBI:17754
propane-1,2,3-triol
C(C(CO)O)O
Viscosity 1069 mPa.s (20°C)
Specific Gravity 1.261
Boiling Point 290.0°C
Refractive Index 1.4716
Packaging Glass bottle
Flash Point 160°C
Infrared Spectrum Authentic
Color Undesignated
Density 1.2610g/mL
Quantity 2.5L
Melting Point 18.0°C
Assay Percent Range 99% min. (GC)
Linear Formula HOCH2CH(OH)CH2OH
Beilstein 01,502
Merck Index 15,452
Solubility Information Solubility in water: >500g/L (20°C). Other solubilities: soluble in methanol, ethanol and 2-propanol, insoluble in ether, benzene, chloroform
Formula Weight 92.09
Physical Form Viscous Liquid
Percent Purity 99+%
Grade Pure
Chemical Name or Material Glycerol
Show More Show Less

EINECSNumber : 200-289-5