Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Glycerol, puriss., Meets analytical specification of Ph. Eur. BP, USP, FCC., Honeywell™ Riedel-de Haen™

Catalog No. 60044885 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
5 L
4 x 5 L
6 x 1 L
250 kg
Packaging:
Poly Bottle
Poly Drum

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E422, anhydrous, 99.0-101.0% (alkalimetric)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO

Specifications

Melting Point 18°C
Color Colorless
Density 1.2580 to 1.2644g/mL at 20°C, ≥1.257g/mL at 25°C
Boiling Point 290°C
Flash Point 160°C (320°F)
Ignition Residue ≤0.01% (as SO4)
Ignition Point 698°F
Packaging Poly Bottle
Refractive Index n20/D 1.474(lit.)
Linear Formula HOCH2CH(OH)CH2OH
Quantity 1 L
UN Number NONH for all modes of transport
Beilstein 635685
Vapor Density 3.1 (vs air)
Vapor Pressure 1mmHg (20°C)
Formula Weight 92.09g/mol
Percent Purity 99.0 to 101.0% (calc. on H2O free substance)
Grade puriss.
Physical Form Liquid
Chemical Name or Material Glycerol, Meets analytical specification of Ph. Eur. BP, USP, FCC.
Show More Show Less
EINECSNumber 200-289-5
RTECSNumber MA8050000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.